Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Methyl-1,2-phenylenediamine Dihydrochloride 98.0+%, TCI America™
SDP

Catalog No. M175425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M175425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M175425G Supplier TCI America Supplier No. M175425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 25148-68-9
Molecular Formula C7H12Cl2N2
Molecular Weight (g/mol) 195.087
MDL Number MFCD00042021
InChI Key DKEONVNYXODZRQ-UHFFFAOYSA-N
PubChem CID 91296
IUPAC Name 2-N-methylbenzene-1,2-diamine;dihydrochloride
SMILES CNC1=CC=CC=C1N.Cl.Cl

Specifications

Melting Point 192°C
Quantity 25 g
UN Number 2811
Formula Weight 195.09
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-Methyl-1,2-phenylenediamine Dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.