accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaH3COOO

CAS RN 2516-93-0

IUPAC Name: sodium 2-butoxyacetate
Synonyms: sodium 2-butoxyacetate
Molecular Weight (g/mol): 154.14
Molecular Formula: C6H11NaO3
InChi Key: VFOVBGNGALJINB-UHFFFAOYSA-M
SMILES: [Na+].CCCCOCC([O-])=O
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)
  • (1)