accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SnH3CCH3NSCH3

CAS RN 251635-59-3

IUPAC Name: 4-methyl-2-(tributylstannyl)-1,3-thiazole
Synonyms: 4-methyl-2-(tributylstannyl)-1,3-thiazole
Molecular Weight (g/mol): 388.2
Molecular Formula: C16H31NSSn
InChi Key: PZPGNMUMILSRSX-UHFFFAOYSA-N
SMILES: CCCC[Sn](CCCC)(CCCC)C1=NC(C)=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results