Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tributyl(4-methyl-1,3-thiazol-2-yl)stannane, Thermo Scientific™

Catalog No. CC75541DA
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC75541DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC75541DA Supplier Thermo Scientific Chemicals Supplier No. CC75541DA

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 251635-59-3
Molecular Formula C16H31NSSn
Molecular Weight (g/mol) 388.201
InChI Key PZPGNMUMILSRSX-UHFFFAOYSA-N
Synonym 4-methyl-2-tributylstannyl thiazole, 4-methyl-2-tributylstannyl-1,3-thiazole, 2-tributylstannyl-4-methylthiazole, thiazole, 4-methyl-2-tributylstannyl, 4-methyl-2-tributylstannanyl-thiazole, acmc-209gho, 4-methyl-2-tributylstannylthiazole, 4-methyl-2-tributylstannanylthiazole, 4-methyl-2-tributylstannyl-thiazole, 4-methyl-2-tributylstannanyl-1,3-thiazole
PubChem CID 16427130
IUPAC Name tributyl-(4-methyl-1,3-thiazol-2-yl)stannane
SMILES CCCC[Sn](CCCC)(CCCC)C1=NC(=CS1)C

Specifications

Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 388.18
Percent Purity 90%
Chemical Name or Material tributyl(4-methyl-1, 3-thiazol-2-yl)stannane
Hazard Category Danger
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.