accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 25173-72-2

IUPAC Name: 3-(3,4,5-trimethoxyphenyl)propanoic acid
Synonyms: 3-(3,4,5-trimethoxyphenyl)propanoic acid
Molecular Weight (g/mol): 240.26
Molecular Formula: C12H16O5
InChi Key: ZCYXGVJUZBKJAI-UHFFFAOYSA-N
SMILES: COC1=CC(CCC(O)=O)=CC(OC)=C1OC
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results