Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(3,4,5-Trimethoxyphenyl)propionic acid, 98%

Catalog No. AC189160250
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC189160250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC189160250 Supplier Thermo Scientific Chemicals Supplier No. 189160250
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 25173-72-2 | C12H16O5 | 240.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25173-72-2
Molecular Formula C12H16O5
Molecular Weight (g/mol) 240.25
MDL Number MFCD00002775
InChI Key ZCYXGVJUZBKJAI-UHFFFAOYSA-N
Synonym 3-3,4,5-trimethoxyphenyl propanoic acid, 3-3,4,5-trimethoxyphenyl propionic acid, benzenepropanoic acid, 3,4,5-trimethoxy, 3,4,5-trimethoxyhydrocinnamic acid, 3,4,5-trimethoxydihydrocinnamic acid, 3,4,5-trimethoxyphenylpropionic acid, 3,4,5-trimethoxybenzenepropanoic acid, 3,4,5-trimethoxy-benzenepropanoic acid, acmc-1cazt, maybridge1_005958
PubChem CID 64860
ChEBI CHEBI:583580
IUPAC Name 3-(3,4,5-trimethoxyphenyl)propanoic acid
SMILES COC1=CC(=CC(=C1OC)OC)CCC(=O)O

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point 100.0°C to 105.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula (CH3O)3C6H2CH2CH2CO2H
Quantity 25 g
Beilstein 10, III, 2120
Formula Weight 240.25
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 3-(3, 4, 5-Trimethoxyphenyl)propionic acid, 98+%
Show More Show Less
Hazard Category GHS Signal Word: Warning
EINECSNumber 246-706-4

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.