accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COHNHOOOH

CAS RN 252049-02-8

IUPAC Name: (9H-fluoren-9-yl)methyl N-(1,3-dihydroxybutan-2-yl)carbamate
Synonyms: 9H-fluoren-9-ylmethyl N-(1,3-dihydroxybutan-2-yl)carbamate
Molecular Weight (g/mol): 327.38
Molecular Formula: C19H21NO4
InChi Key: YOKDHMTZJSRRIQ-UHFFFAOYNA-N
SMILES: CC(O)C(CO)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results