accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 252049-11-9

OOOHOONHHNN(S)

CAS RN 252049-11-9

IUPAC Name: (2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-6-[(pyridin-3-yl)formamido]hexanoic acid
Synonyms: (2S)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-6-(pyridin-3-ylformamido)hexanoic acid
Molecular Weight (g/mol): 473.53
Molecular Formula: C27H27N3O5
InChi Key: CRRQIRUSEYKWPF-DEOSSOPVSA-N
SMILES: OC(=O)[C@H](CCCCNC(=O)C1=CC=CN=C1)NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)