accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SiCH3H3COSOOFFF

CAS RN 252054-88-9

IUPAC Name: 1-(trimethylsilyl)naphthalen-2-yl trifluoromethanesulfonate
Synonyms: 1-(trimethylsilyl)naphthalen-2-yl trifluoromethanesulfonate
Molecular Weight (g/mol): 348.41
Molecular Formula: C14H15F3O3SSi
InChi Key: DHFMFICBVHCWGQ-UHFFFAOYSA-N
SMILES: C[Si](C)(C)C1=C2C=CC=CC2=CC=C1OS(=O)(=O)C(F)(F)F
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results