accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 25234-60-0

IUPAC Name: [2-(dodecanoyloxy)ethyl]trimethylazanium chloride
Synonyms: [2-(dodecanoyloxy)ethyl]trimethylazanium chloride
Molecular Weight (g/mol): 321.93
Molecular Formula: C17H36ClNO2
InChi Key: BBJUKVPDIPYNBS-UHFFFAOYSA-M
SMILES: [Cl-].CCCCCCCCCCCC(=O)OCC[N+](C)(C)C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results