accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OHNSH

CAS RN 2524-78-9

IUPAC Name: N-[3-(sulfanylmethyl)phenyl]acetamide
Synonyms: N-[3-(sulfanylmethyl)phenyl]acetamide
Molecular Weight (g/mol): 181.25
Molecular Formula: C9H11NOS
InChi Key: XKSHGBQHPUBJLB-UHFFFAOYSA-N
SMILES: CC(=O)NC1=CC=CC(CS)=C1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results