Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3'-(Methylthio)acetanilide, TCI America™
SDP

Catalog No. M167125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M167125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M167125G Supplier TCI America Supplier No. M167125G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2524-78-9
Molecular Formula C9H11NOS
Molecular Weight (g/mol) 181.253
InChI Key DFRHNWNBWOIBRW-UHFFFAOYSA-N
Synonym 3-Acetamidothioanisole
PubChem CID 291818
IUPAC Name N-(3-methylsulfanylphenyl)acetamide
SMILES CC(=O)NC1=CC(=CC=C1)SC

Specifications

Quantity 25 g
Formula Weight 181.25
Physical Form Crystalline Powder
Chemical Name or Material 3′-(Methylthio)acetanilide
RTECSNumber BW7025000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.