accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOCH3OCH3OH

CAS RN 25265-71-8

IUPAC Name: 1-(1-hydroxypropoxy)propan-1-ol
Synonyms: dipropylene glycol
Molecular Weight (g/mol): 134.18
Molecular Formula: C6H14O3
InChi Key: QYSXYAURTRCDJU-UHFFFAOYNA-N
SMILES: CCC(O)OC(O)CC
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (4)
  • (3)
Boiling Point 
  • (4)
  • (2)
  • (2)
Physical Form 
  • (4)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (8)
  • (4)
  • (4)
  • (1)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (4)
  • (3)
  • (4)
  • (2)
  • (2)
  • (4)
  • (4)
  • (4)
  • (1)