accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3CH3H3C

CAS RN 25265-78-5

IUPAC Name: 1,2,3,5-tetrapropylbenzene
Synonyms: 1,2,3,5-tetrapropylbenzene
Molecular Weight (g/mol): 246.44
Molecular Formula: C18H30
InChi Key: NYIQECIYDUSNRC-UHFFFAOYSA-N
SMILES: CCCC1=CC(CCC)=C(CCC)C(CCC)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results