Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dodecylbenzene (hard type) (mixture of branched chain isomers), TCI America™
SDP

Catalog No. D098825ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D098825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D098825ML Supplier TCI America Supplier No. D098825ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 25265-78-5
Molecular Formula C18H30
Molecular Weight (g/mol) 246.438
MDL Number MFCD00008974
InChI Key KWKXNDCHNDYVRT-UHFFFAOYSA-N
Synonym Alkylbenzene, Laurylbenzene
PubChem CID 31237
IUPAC Name dodecylbenzene
SMILES CCCCCCCCCCCCC1=CC=CC=C1

Specifications

Color Colorless
Quantity 25 mL
Formula Weight 246.44
Physical Form Liquid
Chemical Name or Material Dodecylbenzene (hard type) (mixture of branched chain isomers)
EINECSNumber (3)-0022
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.