accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNOOH2NHNNH2

CAS RN 252944-01-7

IUPAC Name: N2-(2-aminoethyl)-5-nitropyridine-2,6-diamine
Synonyms: N2-(2-aminoethyl)-5-nitropyridine-2,6-diamine
Molecular Weight (g/mol): 197.2
Molecular Formula: C7H11N5O2
InChi Key: JNXKEBLSRILDQM-UHFFFAOYSA-N
SMILES: NCCNC1=NC(N)=C(C=C1)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results