Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Diamino-N2-(2-aminoethyl)-5-nitropyridine 98.0+%, TCI America™
SDP

Catalog No. D524550MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
50 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D524550MG 50 mg
D5245250MG 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D524550MG Supplier TCI America Supplier No. D524550MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 252944-01-7
Molecular Formula C7H11N5O2
Molecular Weight (g/mol) 197.198
MDL Number MFCD18379166
InChI Key JNXKEBLSRILDQM-UHFFFAOYSA-N
Synonym 2,6-Diamino-N6-(2-aminoethyl)-3-nitropyridine, N2-(2-Aminoethyl)-5-nitropyridine-2,6-diamine
PubChem CID 22142380
IUPAC Name 6-N-(2-aminoethyl)-3-nitropyridine-2,6-diamine
SMILES C1=CC(=NC(=C1[N+](=O)[O-])N)NCCN

Specifications

Color White-Yellow
Quantity 50 mg
Formula Weight 197.20
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Diamino-N2-(2-aminoethyl)-5-nitropyridine
TSCA No
Recommended Storage Freezer (-20 °C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.