accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 25312-65-6

IUPAC Name: 4-[(1R,3aS,3bR,9aS,9bS,11aR)-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl]pentanoic acid
Synonyms: 4-[(1R,3aS,3bR,9aS,9bS,11aR)-9a,11a-dimethyl-tetradecahydro-1H-cyclopenta[a]phenanthren-1-yl]pentanoic acid
Molecular Weight (g/mol): 360.58
Molecular Formula: C24H40O2
InChi Key: RPKLZQLYODPWTM-FOEWTTMCSA-N
SMILES: CC(CCC(O)=O)[C@H]1CC[C@H]2[C@@H]3CCC4CCCC[C@]4(C)[C@H]3CC[C@]12C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results