Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cholanic Acid, MP Biomedicals

Catalog No. ICN205648 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN205648 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN205648 Supplier MP Biomedicals, Inc Supplier No. 0520564810

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 25312-65-6
Molecular Formula C24H40O2
Molecular Weight (g/mol) 360.582
InChI Key RPKLZQLYODPWTM-KBMWBBLPSA-N
Synonym cholan-24-oic acid, cholanic acid, cholan-24-oic acid, 5.beta., 5.beta.-cholanic acid, 5.beta.-cholanoic acid, 5.beta.-cholan-24-oic acid, androstane-17-butanoic acid, .gamma.-methyl-, 5.beta.,17.beta.
PubChem CID 9548797
ChEBI CHEBI:36237
IUPAC Name (4R)-4-[(8R,9S,10S,13R,14S,17R)-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoic acid
SMILES CC(CCC(=O)O)C1CCC2C1(CCC3C2CCC4C3(CCCC4)C)C

Specifications

Quantity 100 mg
Formula Weight 360.59
Chemical Name or Material Cholanic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.