accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NHNNO(E/Z)

CAS RN 253124-08-2

IUPAC Name: 2-propyl-4-[2-(pyridin-4-yl)hydrazin-1-ylidene]cyclohexa-2,5-dien-1-one
Synonyms: 2-propyl-4-[2-(pyridin-4-yl)hydrazin-1-ylidene]cyclohexa-2,5-dien-1-one
Molecular Weight (g/mol): 241.29
Molecular Formula: C14H15N3O
InChi Key: PHBLNRCSUHMCPW-UHFFFAOYSA-N
SMILES: CCCC1=CC(C=CC1=O)=NNC1=CC=NC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results