Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Propyl-4-(4-pyridylazo)phenol 98.0+%, TCI America™
SDP

Catalog No. P14425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P14425G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P14425G Supplier TCI America Supplier No. P14425G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 253124-08-2
Molecular Formula C14H15N3O
Molecular Weight (g/mol) 241.29
MDL Number MFCD06797155
InChI Key PHBLNRCSUHMCPW-UHFFFAOYSA-N
PubChem CID 131674912
IUPAC Name 2-propyl-4-[2-(pyridin-4-yl)hydrazin-1-ylidene]cyclohexa-2,5-dien-1-one
SMILES CCCC1=CC(C=CC1=O)=NNC1=CC=NC=C1

Specifications

Melting Point 177°C
Color Red-Yellow
Quantity 5 g
Formula Weight 241.29
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Propyl-4-(4-pyridylazo)phenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.