accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrCH3NH3CCH3

CAS RN 25316-59-0

IUPAC Name: benzyltributylazanium bromide
Synonyms: benzyltributylazanium bromide
Molecular Weight (g/mol): 356.39
Molecular Formula: C19H34BrN
InChi Key: UDYGXWPMSJPFDG-UHFFFAOYSA-M
SMILES: [Br-].CCCC[N+](CCCC)(CCCC)CC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
  • (5)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (5)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (2)