accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONNNOOH

CAS RN 253315-05-8

IUPAC Name: 2-(morpholin-4-yl)pyrimidine-5-carboxylic acid
Synonyms: 2-(morpholin-4-yl)pyrimidine-5-carboxylic acid
Molecular Weight (g/mol): 209.21
Molecular Formula: C9H11N3O3
InChi Key: WMPUFHIOPXMZFI-UHFFFAOYSA-N
SMILES: OC(=O)C1=CN=C(N=C1)N1CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results