accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNNOOH

CAS RN 253315-22-9

IUPAC Name: 6-(1H-pyrazol-1-yl)pyridine-3-carboxylic acid
Synonyms: 6-(pyrazol-1-yl)pyridine-3-carboxylic acid
Molecular Weight (g/mol): 189.17
Molecular Formula: C9H7N3O2
InChi Key: QWFKXYLAKWFQLF-UHFFFAOYSA-N
SMILES: OC(=O)C1=CN=C(C=C1)N1C=CC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results