accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 25343-91-3

IUPAC Name: 4-methyl-1λ⁶-thiomorpholine-1,1-dione
Synonyms: 4-methyl-1λ⁶-thiomorpholine-1,1-dione
Molecular Weight (g/mol): 149.21
Molecular Formula: C5H11NO2S
InChi Key: ULZCOWMSBOJCLT-UHFFFAOYSA-N
SMILES: CN1CCS(=O)(=O)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results