accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2538-85-4

IUPAC Name: sodium (4E)-4-[2-(2-hydroxynaphthalen-1-yl)hydrazin-1-ylidene]-3-oxo-3,4-dihydronaphthalene-1-sulfonate
Synonyms: sodium (4E)-4-[2-(2-hydroxynaphthalen-1-yl)hydrazin-1-ylidene]-3-oxonaphthalene-1-sulfonate
Molecular Weight (g/mol): 416.38
Molecular Formula: C20H13N2NaO5S
InChi Key: YJUGDMOGPJJWDG-QPNALZDCSA-M
SMILES: [Na+].OC1=CC=C2C=CC=CC2=C1N\N=C1\C(=O)C=C(C2=CC=CC=C12)S([O-])(=O)=O
Molecular Weight (g/mol) 
  • (1)
  • (6)
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (3)
  • (1)
  • (5)
  • (1)
Color 
  • (1)
  • (2)
  • (1)
  • (3)
  • (3)
Grade 
  • (3)
  • (2)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (1)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (6)
  • (2)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (5)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (3)
  • (3)
  • (2)
  • (4)