Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mordant Black 17 [Indicator for Complexometry], TCI America™
SDP

Catalog No. A058125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A058125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A058125G Supplier TCI America Supplier No. A058125G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2538-85-4
Molecular Formula C20H13N2NaO5S
Molecular Weight (g/mol) 416.383
MDL Number MFCD00004069
InChI Key YJUGDMOGPJJWDG-UHFFFAOYSA-M
Synonym Calcon, Eriochrome Blue Black R, 1-(2-Hydroxy-1-naphthylazo)-2-naphthol-4-sulfonic Acid Sodium Salt, Sunchromine Blue Black R
PubChem CID 23702480
IUPAC Name sodium;4-[(2-hydroxynaphthalen-1-yl)hydrazinylidene]-3-oxonaphthalene-1-sulfonate
SMILES C1=CC=C2C(=C1)C=CC(=C2NN=C3C4=CC=CC=C4C(=CC3=O)S(=O)(=O)[O-])O.[Na+]

Specifications

Color Black/Purple
Quantity 25 g
Physical Form Crystal/Powder
Formula Weight 416.38
Chemical Name or Material Mordant Black 17 [Indicator for Complexometry]
EINECSNumber (5)-2116
RTECSNumber QK2200000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.