accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2OOHOO

CAS RN 25395-22-6

IUPAC Name: 2-(2-carbamoylphenoxy)acetic acid
Synonyms: salicylamide O-acetic acid
Molecular Weight (g/mol): 195.17
Molecular Formula: C9H9NO4
InChi Key: RLISWLLILOTWGG-UHFFFAOYSA-N
SMILES: NC(=O)C1=CC=CC=C1OCC(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results