Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(2-Carbamoylphenoxy)acetic Acid 99.0+%, TCI America™
SDP

Catalog No. C057825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C057825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C057825G Supplier TCI America Supplier No. C057825G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 25395-22-6
Molecular Formula C9H9NO4
Molecular Weight (g/mol) 195.17
MDL Number MFCD00025472
InChI Key RLISWLLILOTWGG-UHFFFAOYSA-N
Synonym 2-(Carboxymethoxy)benzamide, Salicylamide O-Acetic Acid
PubChem CID 93086
IUPAC Name 2-(2-carbamoylphenoxy)acetic acid
SMILES NC(=O)C1=CC=CC=C1OCC(O)=O

Specifications

Melting Point 222°C
Color White
Quantity 25 g
Formula Weight 195.17
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (2-Carbamoylphenoxy)acetic Acid
RTECSNumber AF7893000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.