accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2ONaNH2(S)IOIOHIIOO

CAS RN 25416-65-3

IUPAC Name: sodium (2S)-2-amino-3-[4-(4-hydroxy-3,5-diiodophenoxy)-3,5-diiodophenyl]propanoate hydrate
Synonyms: sodium levothyroxine(1-) hydrate
Molecular Weight (g/mol): 816.87
Molecular Formula: C15H12I4NNaO5
InChi Key: ANMYAHDLKVNJJO-LTCKWSDVSA-M
SMILES: O.[Na+].N[C@@H](CC1=CC(I)=C(OC2=CC(I)=C(O)C(I)=C2)C(I)=C1)C([O-])=O
Grade 
  • (6)
Molecular Weight (g/mol) 
  • (5)
  • (1)
Percent Purity 
  • (5)
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (6)
  • (5)
  • (1)
  • (1)
  • (6)
  • (5)
  • (1)
  • (5)
  • (1)
  • (1)