Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Levothyroxine Sodium, USP, 97-103%, Spectrum™ Chemical
SDP

Catalog No. 18603818 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-603-818 25 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-603-818 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. T128125GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

T1281, 25416-65-3, C15H10I4NO4Na·xH2O

Spectrum™ Chemical Levothyroxine Sodium, USP is a synthetic thyroid hormone that is used to treat thyroid hormone deficiency or to prevent thyroid cancer. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 25416-65-3
Molecular Formula C15H12I4NNaO5
Molecular Weight (g/mol) 816.87
MDL Number MFCD06408007,MFCD00002596,MFCD00149110
InChI Key ANMYAHDLKVNJJO-LTCKWSDVSA-M
IUPAC Name sodium (2S)-2-amino-3-[4-(4-hydroxy-3,5-diiodophenoxy)-3,5-diiodophenyl]propanoate hydrate
SMILES O.[Na+].N[C@@H](CC1=CC(I)=C(OC2=CC(I)=C(O)C(I)=C2)C(I)=C1)C([O-])=O

Specifications

CAS Max % 100%
Packaging Amber Glass Bottle
Quantity 25 g
Specific Rotation -5° to -6°
Percent Purity 97 to 103%
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.