CAS RN 2544-05-0
IUPAC Name:
2-chloro-3-methoxypropanoic acid
Synonyms:
2-chloro-3-methoxypropanoic acid
Molecular Weight (g/mol):
138.55
Molecular Formula:
C4H7ClO3
InChi Key:
SSWGTERYCJZJPQ-UHFFFAOYNA-N
SMILES:
COCC(Cl)C(O)=O
1
–
1
of
1
results
1
2-Chloro-3-methoxypropionic Acid 98.0+%, TCI America™
CAS: 2544-05-0 Molecular Formula: C4H7ClO3 Molecular Weight (g/mol): 138.547 MDL Number: MFCD03093624 InChI Key: SSWGTERYCJZJPQ-UHFFFAOYSA-N PubChem CID: 14269106 IUPAC Name: 2-chloro-3-methoxypropanoic acid SMILES: COCC(C(=O)O)Cl