Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-3-methoxypropionic Acid 98.0+%, TCI America™
SDP

Catalog No. C16885G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C16885G 5 g
C168825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C16885G Supplier TCI America Supplier No. C16885G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2544-05-0
Molecular Formula C4H7ClO3
Molecular Weight (g/mol) 138.547
MDL Number MFCD03093624
InChI Key SSWGTERYCJZJPQ-UHFFFAOYSA-N
PubChem CID 14269106
IUPAC Name 2-chloro-3-methoxypropanoic acid
SMILES COCC(C(=O)O)Cl

Specifications

Color Yellow
Boiling Point 155°C
Quantity 5 g
UN Number 3265
Formula Weight 138.55
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Chloro-3-methoxypropionic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.