accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2SNH

CAS RN 25444-82-0

IUPAC Name: (adamantan-1-yl)thiourea
Synonyms: adamantan-1-ylthiourea
Molecular Weight (g/mol): 210.34
Molecular Formula: C11H18N2S
InChi Key: LRWQENBAFMBIJR-UHFFFAOYSA-N
SMILES: NC(=S)NC12CC3CC(CC(C3)C1)C2
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results