Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Adamantylthiourea 97.0+%, TCI America™
SDP

Catalog No. A14035G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A14035G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A14035G Supplier TCI America Supplier No. A14035G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 25444-82-0
Molecular Formula C11H18N2S
Molecular Weight (g/mol) 210.339
MDL Number MFCD01321124
InChI Key LRWQENBAFMBIJR-UHFFFAOYSA-N
PubChem CID 796429
IUPAC Name 1-adamantylthiourea
SMILES C1C2CC3CC1CC(C2)(C3)NC(=S)N

Specifications

Melting Point 167°C
Color White-Yellow
Quantity 5 g
Formula Weight 210.34
Percent Purity ≥97.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 1-Adamantylthiourea
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.