accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2COOO

CAS RN 25447-83-0

IUPAC Name: 3-(dec-9-en-1-yl)oxolane-2,5-dione
Synonyms: 3-(dec-9-en-1-yl)oxolane-2,5-dione
Molecular Weight (g/mol): 238.33
Molecular Formula: C14H22O3
InChi Key: JVKGGERSFUBGEE-UHFFFAOYNA-N
SMILES: C=CCCCCCCCCC1CC(=O)OC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results