Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Decenylsuccinic Anhydride 95.0+%, TCI America™
SDP

Catalog No. D003025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D003025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D003025G Supplier TCI America Supplier No. D003025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 25447-83-0
Molecular Formula C14H22O3
Molecular Weight (g/mol) 238.327
MDL Number MFCD00014547
InChI Key UWERUIGPWOVNGG-MDZDMXLPSA-N
PubChem CID 5354807
IUPAC Name 3-[(E)-dec-1-enyl]oxolane-2,5-dione
SMILES CCCCCCCCC=CC1CC(=O)OC1=O

Specifications

Color Yellow
Quantity 25 g
Formula Weight 238.33
Percent Purity ≥95.0% (T)
Physical Form Liquid
Chemical Name or Material Decenylsuccinic Anhydride
EINECSNumber (2)-0852
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.