accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClClClSClOO

CAS RN 2547-61-7

IUPAC Name: trichloromethanesulfonyl chloride
Synonyms: trichloromethanesulfonyl chloride
Molecular Weight (g/mol): 217.87
Molecular Formula: CCl4O2S
InChi Key: ZCPSWAFANXCCOT-UHFFFAOYSA-N
SMILES: ClC(Cl)(Cl)S(Cl)(=O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)