Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trichloromethanesulfonyl Chloride 95.0+%, TCI America™
SDP

Catalog No. T06115G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T06115G 5 g
T061125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T06115G Supplier TCI America Supplier No. T06115G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2547-61-7
Molecular Formula CCl4O2S
Molecular Weight (g/mol) 217.869
MDL Number MFCD00007452
InChI Key ZCPSWAFANXCCOT-UHFFFAOYSA-N
PubChem CID 75684
IUPAC Name trichloromethanesulfonyl chloride
SMILES C(S(=O)(=O)Cl)(Cl)(Cl)Cl

Specifications

Melting Point 141°C
Color White-Yellow
Quantity 5 g
UN Number 3261
Formula Weight 217.87
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Trichloromethanesulfonyl Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.