accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2549-87-3

IUPAC Name: 2-benzoyl-5-(prop-2-en-1-yloxy)phenol
Synonyms: 2-benzoyl-5-(prop-2-en-1-yloxy)phenol
Molecular Weight (g/mol): 254.29
Molecular Formula: C16H14O3
InChi Key: GVZIBGFELWPEOC-UHFFFAOYSA-N
SMILES: OC1=CC(OCC=C)=CC=C1C(=O)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results