Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Allyloxy-2-hydroxybenzophenone 98.0+%, TCI America™
SDP

Catalog No. A234225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A234225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A234225G Supplier TCI America Supplier No. A234225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2549-87-3
Molecular Formula C16H14O3
Molecular Weight (g/mol) 254.29
MDL Number MFCD00191713
InChI Key GVZIBGFELWPEOC-UHFFFAOYSA-N
PubChem CID 75689
IUPAC Name 2-benzoyl-5-(prop-2-en-1-yloxy)phenol
SMILES OC1=CC(OCC=C)=CC=C1C(=O)C1=CC=CC=C1

Specifications

Melting Point 68°C
Color Yellow
Boiling Point 178°C
Quantity 25 g
Formula Weight 254.29
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Allyloxy-2-hydroxybenzophenone
EINECSNumber (4)-0135
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.