accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 25496-72-4

IUPAC Name: 2,3-dihydroxypropyl (9Z)-octadec-9-enoate
Synonyms: oleoyl glycerol
Molecular Weight (g/mol): 356.55
Molecular Formula: C21H40O4
InChi Key: RZRNAYUHWVFMIP-KTKRTIGZNA-N
SMILES: CCCCCCCC\C=C/CCCCCCCC(=O)OCC(O)CO
Grade 
  • (1)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (4)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (1)
  • (4)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (4)