Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glyceryl Monooleate, MP Biomedicals

Catalog No. ICN211013 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN211013 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN211013 Supplier MP Biomedicals, Inc Supplier No. 0521101380
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 25496-72-4
Molecular Formula C21H40O4
Molecular Weight (g/mol) 356.55
MDL Number MFCD00067258,MFCD00042735,MFCD00069635
InChI Key RZRNAYUHWVFMIP-KTKRTIGZNA-N
Synonym monoelaidin, 1-monoelaidin, dl-alpha-monoolein, 2,3-dihydroxypropyl elaidate, glycerol monooleate, 9-octadecenoic acid, 2,3-dihydroxypropyl ester, e, 2,3-dihydroxypropyl 9e-octadec-9-enoate, 1-elaidoylglycerol, arlacel tm 186, glycerol-1-monooleate
PubChem CID 5364833
IUPAC Name 2,3-dihydroxypropyl (9Z)-octadec-9-enoate
SMILES CCCCCCCC\C=C/CCCCCCCC(=O)OCC(O)CO

Specifications

Quantity 100 g
Formula Weight 356.55
Chemical Name or Material Glyceryl Monooleate
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.