accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 25560-00-3

IUPAC Name: 3-(3-methylpiperidin-1-yl)propan-1-amine
Synonyms: 3-(3-methylpiperidin-1-yl)propan-1-amine
Molecular Weight (g/mol): 156.27
Molecular Formula: C9H20N2
InChi Key: ADIQUBKLXXCKLT-UHFFFAOYNA-N
SMILES: CC1CCCN(CCCN)C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results