accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 25564-22-1

IUPAC Name: 2-pentylcyclopent-2-en-1-one
Synonyms: amylcyclopentenone
Molecular Weight (g/mol): 152.24
Molecular Formula: C10H16O
InChi Key: ILHZVKAXFCDFMT-UHFFFAOYSA-N
SMILES: CCCCCC1=CCCC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results