Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Pentyl-2-cyclopenten-1-one 95.0+%, TCI America™
SDP

Catalog No. P114225ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P114225ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P114225ML Supplier TCI America Supplier No. P114225ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 25564-22-1
Molecular Formula C10H16O
Molecular Weight (g/mol) 152.237
MDL Number MFCD00036631
InChI Key ILHZVKAXFCDFMT-UHFFFAOYSA-N
PubChem CID 117549
IUPAC Name 2-pentylcyclopent-2-en-1-one
SMILES CCCCCC1=CCCC1=O

Specifications

Color Yellow
Boiling Point 115°C
Quantity 25 mL
Formula Weight 152.24
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Pentyl-2-cyclopenten-1-one
EINECSNumber (3)-3598
RTECSNumber GY7310000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.