accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHHOHOOOHO

CAS RN 25569-53-3

IUPAC Name: butanedioic acid; ethane-1,2-diol
Synonyms: poly(ethylene succinate)
Molecular Weight (g/mol): 180.16
Molecular Formula: C6H12O6
InChi Key: VJVOPINBJQWMNY-UHFFFAOYSA-N
SMILES: OCCO.OC(=O)CCC(O)=O
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results