Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poly(ethylene Glycol Succinate), TCI America™
SDP

Catalog No. P046725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P046725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P046725G Supplier TCI America Supplier No. P046725G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 25569-53-3
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.156
MDL Number MFCD00084421
InChI Key VJVOPINBJQWMNY-UHFFFAOYSA-N
Synonym Ethylene Glycol Succinate Polyester, Succinate Ethylene Glycol Polyester
PubChem CID 33099
IUPAC Name butanedioic acid;ethane-1,2-diol
SMILES C(CC(=O)O)C(=O)O.C(CO)O

Specifications

Color White-Yellow
Quantity 25 g
Physical Form Crystalline Powder
Chemical Name or Material Poly(ethylene Glycol Succinate)
EINECSNumber (7)-0784
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.