CAS RN 25572-95-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Avantor J.T.Baker trans-(1,2-Cyclohexylenedinitrilo)tetraacetic Acid, Monohydrate, BAKER™, J.T. Baker™
CAS: 25572-95-4 Molecular Formula: C14H22N2O8·H2O