Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-(1,2-Cyclohexylenedinitrilo)tetraacetic Acid, Monohydrate, BAKER™, J.T. Baker™

Catalog No. 02004491 Shop All Avantor Products
Change view
Click to view available options
Quantity:
100 g
Packaging:
Wide Mouth Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02-004-491 100 g Wide Mouth Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02-004-491 Supplier Avantor Supplier No. G08305

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

A grade of chemicals for laboratory use

Chemical Identifiers

CAS 25572-95-4
Molecular Formula C14H22N2O8·H2O

Specifications

Packaging Wide Mouth Amber Glass Bottle
Quantity 100 g
Formula Weight 364.35
Chemical Name or Material trans-(1,2-Cyclohexylenedinitrilo)tetraacetic Acid, Monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.